2-(Trifluoromethyl)-4-chloro-1H-benzimidazole
Supplier: Apollo Scientific
|
Warning
|
PC32931-1G
PC32931-250MG
APOSPC32931-1GEA
575
EUR
APOSPC32931-1G
APOSPC32931-250MG
2-(Trifluoromethyl)-4-chloro-1H-benzimidazole
2-(Trifluoromethyl)-4-chloro-1H-benzimidazole
Formula:
C₈H₄ClF₃N₂ MW: 220,579 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01658394 CAS Number: 2338-31-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...