2-Bromo-4-fluoro-1-cyclopropylbenzene

Supplier: Apollo Scientific
Warning

PC32944-1G PC32944-250MG
APOSPC32944-1GEA 473 EUR
APOSPC32944-1G APOSPC32944-250MG
2-Bromo-4-fluoro-1-cyclopropylbenzene
2-Bromo-4-fluoro-1-cyclopropylbenzene
Formula: C₉H₈BrF
MW: 215.062 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21100009
CAS Number: 1353855-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR