2-Bromo-6-fluoro-1-cyclopropylbenzene
Supplier: Apollo Scientific
|
Warning
|
PC32946-1G
PC32946-250MG
APOSPC32946-1GEA
105
EUR
APOSPC32946-1G
APOSPC32946-250MG
2-Bromo-6-fluoro-1-cyclopropylbenzene
2-Bromo-6-fluoro-1-cyclopropylbenzene
Formula:
C₉H₈BrF MW: 215,062 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24449783 CAS Number: 1415096-07-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...