3-Bromo-2-fluoro-1-cyclopropylbenzene

Supplier: Apollo Scientific
Warning

PC32947-1G PC32947-250MG
APOSPC32947-1GEA 655 EUR
APOSPC32947-1G APOSPC32947-250MG
3-Bromo-2-fluoro-1-cyclopropylbenzene
3-Bromo-2-fluoro-1-cyclopropylbenzene
Formula: C₉H₈BrF
MW: 215,062 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31435752
CAS Number: 2137823-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR