4-Bromo-3-fluoro-1-cyclopropylbenzene
Supplier: Apollo Scientific
|
Warning
|
PC32951-1G
PC32951-250MG
APOSPC32951-1GEA
112
EUR
APOSPC32951-1G
APOSPC32951-250MG
4-Bromo-3-fluoro-1-cyclopropylbenzene
4-Bromo-3-fluoro-1-cyclopropylbenzene
Formula:
C₉H₈BrF MW: 215.062 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD21099975 CAS Number: 1353854-74-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...