5-Bromo-2-fluoro-1-cyclopropylbenzene

Supplier: Apollo Scientific
Warning

PC32952-1G PC32952-250MG
APOSPC32952-1GEA 545 EUR
APOSPC32952-1G APOSPC32952-250MG
5-Bromo-2-fluoro-1-cyclopropylbenzene
5-Bromo-2-fluoro-1-cyclopropylbenzene
Formula: C₉H₈BrF
MW: 215,062 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19439948
CAS Number: 1345035-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR