Perfluorbutanesulphonylamide(N-methyl)acetate 97%

Supplier: Apollo Scientific
Warning

Synonyms: N-Methyl-N-[(1,1,2,2,3,3,4,4,4-nonafluorobutyl)sulphonyl] glycine

PC32961-100MG PC32961-250MG
APOSPC32961-100MGEA 405 EUR
APOSPC32961-100MG APOSPC32961-250MG
Perfluorbutanesulphonylamide(N-methyl)acetate 97%
Perfluorbutanesulphonylamide(N-methyl)acetate
Formula: C₇H₆F₉NO₄S
MW: 371,177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32011191
CAS Number: 159381-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR