4-Acetamido-3-fluorobenzeneboronic acid, pinacol ester

Supplier: Apollo Scientific

Synonyms: N-(2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetamide, N-[2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetamide

PC3443-250MG PC3443-50MG
APOSPC3443-250MGEA 473 EUR
APOSPC3443-250MG APOSPC3443-50MG
4-Acetamido-3-fluorobenzeneboronic acid, pinacol ester
4-Acetamido-3-fluorobenzeneboronic acid, pinacol ester
Formula: C₁₄H₁₉BFNO₃
MW: 279,115 g/mol
Melting Pt: 118…120 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09878424
CAS Number: 269410-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR