7-Fluoroisatin-3-oxime 98%
Supplier: Apollo Scientific
|
Warning
|
PC3583-1G
PC3583-25G
PC3583-5G
APOSPC3583-1GEA
21.9
EUR
APOSPC3583-1G
APOSPC3583-25G
APOSPC3583-5G
7-Fluoroisatin-3-oxime 98%
7-Fluoroisatin-3-oxime
Formula:
C₈H₅FN₂O₂ Melting Pt: 207…208 °C Storage Temperature: Ambient |
MDL Number:
MFCD02951896 CAS Number: 143884-84-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...