n-Butyl pentafluoropropionate 98%

Supplier: Apollo Scientific

PC3771-25G PC3771-10G PC3771-100G
APOSPC3771-25GEA 219 EUR
APOSPC3771-25G APOSPC3771-10G APOSPC3771-100G
n-Butyl pentafluoropropionate 98%
n-Butyl pentafluoropropionate
Formula: C₇H₉F₅O₂
MW: 220,137 g/mol
Boiling Pt: 116…117 °C
Storage Temperature: Ambient
MDL Number: MFCD02093312
CAS Number: 680-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR