Methyl nonafluorobutane-1-sulphonate

Supplier: Apollo Scientific
Danger

Synonyms: Methyl nonaflate, Methyl perfluorobutane-1-sulphonate, Methyl 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulphonate

PC3785J-1G PC3785J-5G
APOSPC3785J-1GEA 256 EUR
APOSPC3785J-1G APOSPC3785J-5G
Methyl nonafluorobutane-1-sulphonate
Methyl nonafluorobutane-1-sulphonate
Formula: C₅H₃F₉O₃S
MW: 314,126 g/mol
Boiling Pt: 155…156 °C
Density: 1,688 g/cm³ (25 °C)
Flash Pt: 50 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00210080
CAS Number: 6401-03-2
UN: 2929
ADR: 6.1+3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR