3-Fluoro-4-methylbenzamide oxime 97%

Supplier: Apollo Scientific

PC3822G-1G PC3822G-5G
APOSPC3822G-1GEA 125 EUR
APOSPC3822G-1G APOSPC3822G-5G
3-Fluoro-4-methylbenzamide oxime 97%
3-Fluoro-4-methylbenzamide oxime
Formula: C₈H₉FN₂O
Melting Pt: 105…107 °C
Storage Temperature: Ambient
MDL Number: MFCD08461808
CAS Number: 238742-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR