7-Benzyloxy-4-(trifluoromethyl)coumarin 97%
Supplier: Apollo Scientific
|
Warning
|
PC3933-1G
PC3933-250MG
APOSPC3933-1GEA
579
EUR
APOSPC3933-1G
APOSPC3933-250MG
7-Benzyloxy-4-(trifluoromethyl)coumarin 97%
7-Benzyloxy-4-(trifluoromethyl)coumarin
Formula:
C₁₇H₁₁F₃O₃ MW: 320,263 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD01564466 CAS Number: 220001-53-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...