Methyl 4-(ethylamino)-6-fluoropyridine-3-carboxylate

Supplier: Apollo Scientific
Warning

PC400034-1G PC400034-250MG
APOSPC400034-1GEA 881 EUR
APOSPC400034-1G APOSPC400034-250MG
Methyl 4-(ethylamino)-6-fluoropyridine-3-carboxylate
Methyl 4-(ethylamino)-6-fluoropyridine-3-carboxylate
Formula: C₉H₁₁FN₂O₂
MW: 198,194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31536749
CAS Number: 2187435-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR