3-Ethoxy-2-fluorobenzaldehyde

Supplier: Apollo Scientific
Warning

PC400046-1G PC400046-250MG
APOSPC400046-1GEA 200 EUR
APOSPC400046-1G APOSPC400046-250MG
3-Ethoxy-2-fluorobenzaldehyde
3-Ethoxy-2-fluorobenzaldehyde
Formula: C₉H₉FO₂
MW: 168,165 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24678362
CAS Number: 1204176-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR