6-Fluoro-2-mercaptoquinazolin-4(3H)-one 95+%

Supplier: Apollo Scientific
Warning

Synonyms: 6-Fluoro-2-thioxo-2,3-dihydroquinazolin-4(1H)-one

PC400048-250MG PC400048-1G
APOSPC400048-250MGEA 65.4 EUR
APOSPC400048-250MG APOSPC400048-1G
6-Fluoro-2-mercaptoquinazolin-4(3H)-one 95+%
6-Fluoro-2-mercaptoquinazolin-4(3H)-one
Formula: C₈H₅FN₂OS
MW: 196,202 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09041760
CAS Number: 69661-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR