tert-Butyl (2-(difluoromethoxy)-5-hydroxypyridin-3-yl)methylcarbamate

Supplier: Apollo Scientific

PC400104-250MG PC400104-500MG
APOSPC400104-250MGEA 1280 EUR
APOSPC400104-250MG APOSPC400104-500MG
tert-Butyl (2-(difluoromethoxy)-5-hydroxypyridin-3-yl)methylcarbamate
tert-Butyl (2-(difluoromethoxy)-5-hydroxypyridin-3-yl)methylcarbamate
Formula: C₁₂H₁₆F₂N₂O₄
MW: 290,263 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31621141
CAS Number: 2231676-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR