2,2,2-Trifluoro-1-(1H-indazol-5-yl)ethanone
Supplier: Apollo Scientific
|
Warning
|
PC400153-1G
PC400153-250MG
APOSPC400153-1GEA
550
EUR
APOSPC400153-1G
APOSPC400153-250MG
2,2,2-Trifluoro-1-(1H-indazol-5-yl)ethanone
2,2,2-Trifluoro-1-(1H-indazol-5-yl)ethanone
Formula:
C₉H₅F₃N₂O MW: 214,144 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD13173240 CAS Number: 403660-48-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...