(5-Bromo-2-(difluoromethoxy)pyridin-3-yl)methanol

Supplier: Apollo Scientific
Warning

PC400163-100MG PC400163-250MG
APOSPC400163-100MGEA 197 EUR
APOSPC400163-100MG APOSPC400163-250MG
(5-Bromo-2-(difluoromethoxy)pyridin-3-yl)methanol
(5-Bromo-2-(difluoromethoxy)pyridin-3-yl)methanol
Formula: C₇H₆BrF₂NO₂
MW: 254,029 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31621145
CAS Number: 2231673-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR