2-(Difluoromethoxy)-5-hydroxypyridine-3-carbonitrile

Supplier: Apollo Scientific
Warning

PC400166-100MG PC400166-250MG
APOSPC400166-100MGEA 230 EUR
APOSPC400166-100MG APOSPC400166-250MG
2-(Difluoromethoxy)-5-hydroxypyridine-3-carbonitrile
2-(Difluoromethoxy)-5-hydroxypyridine-3-carbonitrile
Formula: C₇H₄F₂N₂O₂
MW: 186,116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28760261
CAS Number: 1805653-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR