Methyl 5-amino-3-fluoropicolinate

Supplier: Apollo Scientific
Warning

PC400185-1G PC400185-250MG
APOSPC400185-1GEA 592 EUR
APOSPC400185-1G APOSPC400185-250MG
Methyl 5-amino-3-fluoropicolinate
Methyl 5-amino-3-fluoropicolinate
Formula: C₇H₇FN₂O₂
MW: 170,141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22376791
CAS Number: 1374652-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR