Racemic trans-2-Fluorocyclopropanecarboxylic acid 95+%

Supplier: Apollo Scientific
Warning

Synonyms: trans-2-Fluorocyclopropanecarboxylic acid

PC400541-1G PC400541-10G PC400541-5G
APOSPC400541-1GEA 63.8 EUR
APOSPC400541-1G APOSPC400541-10G APOSPC400541-5G
Racemic trans-2-Fluorocyclopropanecarboxylic acid 95+%
Racemic trans-2-Fluorocyclopropanecarboxylic acid
Formula: C₄H₅FO₂
Storage Temperature: Ambient
MDL Number: MFCD11499062
CAS Number: 130340-04-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR