2-(Chloromethyl)-5-(trifluoromethyl)-1,3,4-oxadiazole

Supplier: Apollo Scientific
Warning

Synonyms: 5-(Chloromethyl)-2-(trifluoromethyl)-1,3,4-oxadiazole

PC400722-1G PC400722-5G
APOSPC400722-1GEA 134 EUR
APOSPC400722-1G APOSPC400722-5G
2-(Chloromethyl)-5-(trifluoromethyl)-1,3,4-oxadiazole
2-(Chloromethyl)-5-(trifluoromethyl)-1,3,4-oxadiazole
Formula: C₄H₂ClF₃N₂O
Storage Temperature: Ambient
MDL Number: MFCD09699166
CAS Number: 723286-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR