2-(4-Fluorophenyl)-1H-imidazole

Supplier: Apollo Scientific

PC402015-1G PC402015-250MG
APOSPC402015-1GEA 217 EUR
APOSPC402015-1G APOSPC402015-250MG
2-(4-Fluorophenyl)-1H-imidazole
2-(4-Fluorophenyl)-1H-imidazole
Formula: C₉H₇FN₂
Storage Temperature: Ambient
MDL Number: MFCD04114652
CAS Number: 4278-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR