7-Fluoro-4-nitro-1H-indole
Supplier: Apollo Scientific
PC402019-250MG
PC402019-1G
APOSPC402019-250MGEA
502
EUR
APOSPC402019-250MG
APOSPC402019-1G
7-Fluoro-4-nitro-1H-indole
7-Fluoro-4-nitro-1H-indole
Formula:
C₈H₅FN₂O₂ MW: 180,136 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD25460309 CAS Number: 548458-05-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...