Pentafluoro(2,2,3,3-tetrafluoropropoxy)benzene
Supplier: Apollo Scientific
|
Warning
|
PC4037-25G
PC4037-5G
APOSPC4037-25GEA
383
EUR
APOSPC4037-25G
APOSPC4037-5G
Pentafluoro(2,2,3,3-tetrafluoropropoxy)benzene
Pentafluoro(2,2,3,3-tetrafluoropropoxy)benzene
Formula:
C₉H₃F₉O Storage Temperature: Ambient |
MDL Number:
MFCD00155914 CAS Number: 89847-87-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...