4-Fluorobenzoyl isocyanate
Supplier: Apollo Scientific
PC404017-1G
PC404017-250MG
APOSPC404017-1GEA
429
EUR
APOSPC404017-1G
APOSPC404017-250MG
4-Fluorobenzoyl isocyanate
4-Fluorobenzoyl isocyanate
May form small amount of sediment
Formula:
C₈H₄FNO₂ MW: 165,121 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00673066 CAS Number: 18354-35-3 UN: 2206 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...