3-(4-Fluoro-benzyl)-piperidine hydrochloride

Supplier: Apollo Scientific

PC404525-1G PC404525-250MG
APOSPC404525-1GEA 0 EUR
APOSPC404525-1G APOSPC404525-250MG
3-(4-Fluoro-benzyl)-piperidine hydrochloride
3-(4-Fluoro-benzyl)-piperidine hydrochloride
Formula: C₁₂H₁₆FN·ClH
MW: 229,721 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04113623
CAS Number: 745817-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR