3-(4-Fluorophenyl)oxetan-3-amine 95%

Supplier: Apollo Scientific

PC405504-1G PC405504-250MG PC405504-500MG
APOSPC405504-1GEA 1110 EUR
APOSPC405504-1G APOSPC405504-250MG APOSPC405504-500MG
3-(4-Fluorophenyl)oxetan-3-amine 95%
3-(4-Fluorophenyl)oxetan-3-amine
Formula: C₉H₁₀FNO
Storage Temperature: Refrigerator
MDL Number: MFCD18251061
CAS Number: 1260672-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR