1-(4-Fluorophenyl)bicyclo[1.1.1]pentane

Supplier: Apollo Scientific

PC405686-1G PC405686-250MG
APOSPC405686-1GEA 1660 EUR
APOSPC405686-1G APOSPC405686-250MG
1-(4-Fluorophenyl)bicyclo[1.1.1]pentane
1-(4-Fluorophenyl)bicyclo[1.1.1]pentane
Formula: C₁₁H₁₁F
MW: 162,203 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18826721
CAS Number: 271767-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR