(4-(3-Fluorobicyclo[1.1.1]pentan-1-yl)phenyl)methanamine

Supplier: Apollo Scientific

PC405694-1G PC405694-250MG
APOSPC405694-1GEA 2410 EUR
APOSPC405694-1G APOSPC405694-250MG
(4-(3-Fluorobicyclo[1.1.1]pentan-1-yl)phenyl)methanamine
(4-(3-Fluorobicyclo[1.1.1]pentan-1-yl)phenyl)methanamine
Formula: C₁₂H₁₄FN
MW: 191,245 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29037341
CAS Number: 1934501-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR