4-(3-Fluorobicyclo[1.1.1]pentan-1-yl)benzoic acid
Supplier: Apollo Scientific
PC405695-1G
PC405695-500MG
APOSPC405695-1GEA
4080
EUR
APOSPC405695-1G
APOSPC405695-500MG
4-(3-Fluorobicyclo[1.1.1]pentan-1-yl)benzoic acid
4-(3-Fluorobicyclo[1.1.1]pentan-1-yl)benzoic acid
Formula:
C₁₂H₁₁FO₂ MW: 206,213 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29037342 CAS Number: 1980053-59-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...