(2R,3R,4S,5S)-4-Fluorocubane-1-carboxylic acid 95%

Supplier: Apollo Scientific

PC405710-1G PC405710-250MG
APOSPC405710-1GEA 4520 EUR
APOSPC405710-1G APOSPC405710-250MG
(2R,3R,4S,5S)-4-Fluorocubane-1-carboxylic acid 95%
(2R,3R,4S,5S)-4-Fluorocubane-1-carboxylic acid
Formula: C₉H₇FO₂
MW: 166,149 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20645019
CAS Number: 145193-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR