2,2,3,3,4,4-Hexafluorobutyric acid

Supplier: Apollo Scientific
Danger

PC408040-1G PC408040-250MG
APOSPC408040-1GEA 2010 EUR
APOSPC408040-1G APOSPC408040-250MG
2,2,3,3,4,4-Hexafluorobutyric acid
2,2,3,3,4,4-Hexafluorobutyric acid
Formula: C₄H₂F₆O₂
Boiling Pt: 151…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00465636
CAS Number: 679-12-9
UN: 1760
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR