2-Bromo-1,1-difluorocyclopropane
Supplier: Apollo Scientific
|
Warning
|
PC408051-1G
PC408051-500MG
APOSPC408051-1GEA
989
EUR
APOSPC408051-1G
APOSPC408051-500MG
2-Bromo-1,1-difluorocyclopropane
2-Bromo-1,1-difluorocyclopropane
Formula:
C₃H₃BrF₂ MW: 156,957 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD20622605 CAS Number: 51326-64-8 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...