3-Chloro-4-(1,1,2,3,3,3-hexafluoropropoxy)aniline 94%
Supplier: Apollo Scientific
|
Warning
|
PC408133-5G
PC408133-1G
APOSPC408133-5GEA
161
EUR
APOSPC408133-5G
APOSPC408133-1G
3-Chloro-4-(1,1,2,3,3,3-hexafluoropropoxy)aniline 94%
3-Chloro-4-(1,1,2,3,3,3-hexafluoropropoxy)aniline
Formula:
C₉H₆ClF₆NO MW: 293,593 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18641846 CAS Number: 1100832-66-3 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...