Octafluorobut-1-ene

Supplier: Apollo Scientific
Warning

PC408169-25G PC408169-5G
APOSPC408169-25GEA 1140 EUR
APOSPC408169-25G APOSPC408169-5G
Octafluorobut-1-ene
Octafluorobut-1-ene
Formula: C₄F₈
MW: 200.03 g/mol
Boiling Pt: 6 °C
Density: 1.54 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD16619652
CAS Number: 357-26-6
UN: 3163
ADR: 2.2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR