2-Bromo-6-fluorophenylacetic acid

Supplier: Apollo Scientific
Warning

Synonyms: (2-Bromo-6-fluorophenyl)acetic acid

PC408232-10G PC408232-5G
APOSPC408232-10GEA 1020 EUR
APOSPC408232-10G APOSPC408232-5G
2-Bromo-6-fluorophenylacetic acid
2-Bromo-6-fluorophenylacetic acid
Formula: C₈H₆BrFO₂
MW: 233,034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185377
CAS Number: 1214322-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR