2-Fluoro-3-methoxyphenylacetic acid
Supplier: Apollo Scientific
|
Warning
|
PC408262-500MG
PC408262-1G
APOSPC408262-500MGEA
115
EUR
APOSPC408262-500MG
APOSPC408262-1G
2-Fluoro-3-methoxyphenylacetic acid
2-Fluoro-3-methoxyphenylacetic acid
Formula:
C₉H₉FO₃ MW: 184,164 g/mol Melting Pt: 106…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD06800745 CAS Number: 863645-29-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...