2-Fluoro-4-methylcinnamic acid 97%

Supplier: Apollo Scientific
Warning

PC408268-1G PC408268-250MG
APOSPC408268-1GEA 444 EUR
APOSPC408268-1G APOSPC408268-250MG
2-Fluoro-4-methylcinnamic acid 97%
2-Fluoro-4-methylcinnamic acid
Formula: C₁₀H₉FO₂
MW: 180,176 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06205750
CAS Number: 773132-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR