4-Methyl-3-(trifluoromethyl)-1H-pyrazole
Supplier: Apollo Scientific
|
Warning
|
PC411333-1G
PC411333-5G
APOSPC411333-1GEA
65.5
EUR
APOSPC411333-1G
APOSPC411333-5G
4-Methyl-3-(trifluoromethyl)-1H-pyrazole
4-Methyl-3-(trifluoromethyl)-1H-pyrazole
Formula:
C₅H₅F₃N₂ MW: 150,102 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01312008 CAS Number: 153085-14-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...