2-Fluoro-3-hydroxymethylphenylboronic acid 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Fluoro-3-(hydroxymethyl)phenylboronic acid, (2-Fluoro-3-(hydroxymethyl)phenyl)boronic acid
PC412062-1G
PC412062-5G
PC412062-25G
APOSPC412062-1GEA
130
EUR
APOSPC412062-1G
APOSPC412062-5G
APOSPC412062-25G
2-Fluoro-3-hydroxymethylphenylboronic acid 97%
2-Fluoro-3-hydroxymethylphenylboronic acid
Formula:
C₇H₈BFO₃ MW: 169,946 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17926485 CAS Number: 1256355-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...