3,5-Difluoro-4-(hydroxymethyl)phenylboronic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: (3,5-Difluoro-4-(hydroxymethyl)phenyl)boronic acid

PC412112-1G PC412112-250MG
APOSPC412112-1GEA 284 EUR
APOSPC412112-1G APOSPC412112-250MG
3,5-Difluoro-4-(hydroxymethyl)phenylboronic acid 98%
3,5-Difluoro-4-(hydroxymethyl)phenylboronic acid
Formula: C₇H₇BF₂O₃
MW: 187.936 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03095141
CAS Number: 917969-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR