2-Hydroxy-5-(trifluoromethoxy)phenylboronic acid
Supplier: Apollo Scientific
|
Warning
|
PC412497-25G
PC412497-5G
PC412497-1G
APOSPC412497-25GEA
945
EUR
APOSPC412497-25G
APOSPC412497-5G
APOSPC412497-1G
2-Hydroxy-5-(trifluoromethoxy)phenylboronic acid
2-Hydroxy-5-(trifluoromethoxy)phenylboronic acid
Formula:
C₇H₆BF₃O₄ MW: 221,926 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18397559 CAS Number: 1354819-26-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...