7-Fluoro-3-hydroxy-3-phenylindolin-2-one 98%
Supplier: Apollo Scientific
PC421227-100MG
PC421227-250MG
APOSPC421227-100MGEA
857
EUR
APOSPC421227-100MG
APOSPC421227-250MG
7-Fluoro-3-hydroxy-3-phenylindolin-2-one 98%
7-Fluoro-3-hydroxy-3-phenylindolin-2-one
Formula:
C₁₄H₁₀FNO₂ MW: 243,233 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29472335 CAS Number: 1584139-77-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...