Methyl 2-(3,5-difluoro-2-hydroxyphenyl)acetate 95%
Supplier: Apollo Scientific
PC421230-100MG
PC421230-250MG
APOSPC421230-100MGEA
522
EUR
APOSPC421230-100MG
APOSPC421230-250MG
Methyl 2-(3,5-difluoro-2-hydroxyphenyl)acetate 95%
Methyl 2-(3,5-difluoro-2-hydroxyphenyl)acetate
Formula:
C₉H₈F₂O₃ MW: 202,155 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28789151 CAS Number: 209982-92-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...