5-Bromo-4-fluoro-2-hydroxybenzaldehyde 98%
Supplier: Apollo Scientific
|
Warning
|
PC430579-5G
APOSPC430579-5GEA
168
EUR
APOSPC430579-5G
5-Bromo-4-fluoro-2-hydroxybenzaldehyde 98%
5-Bromo-4-fluoro-2-hydroxybenzaldehyde
Formula:
C₇H₄BrFO₂ MW: 219.008 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06739493 CAS Number: 399-00-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...