Heptafluorobenzyl iodide 90%

Supplier: Apollo Scientific
Danger

Synonyms: 1-[Difluoro(iodo)methyl]-2,3,4,5,6-pentafluorobenzene, α,α,2,3,4,5,6-Heptafluoro-α-iodotoluene

PC4389Q-5G PC4389Q-1G
APOSPC4389Q-5GEA 263 EUR
APOSPC4389Q-5G APOSPC4389Q-1G
Heptafluorobenzyl iodide 90%
Heptafluorobenzyl iodide
Formula: C₇F₇I
MW: 343,968 g/mol
Boiling Pt: 153 °C
Density: 2,185 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00153184
CAS Number: 79865-03-5
UN: 1760
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR