3-Fluoro-4-nitrobenzoyl chloride 95%

Supplier: Apollo Scientific
Danger

Synonyms: 4-(Chlorocarbonyl)-2-fluoronitrobenzene, 4-(Chloroformyl)-2-fluoronitrobenzene

PC4402-1G PC4402-5G PC4402-100MG
APOSPC4402-1GEA 176 EUR
APOSPC4402-1G APOSPC4402-5G APOSPC4402-100MG
3-Fluoro-4-nitrobenzoyl chloride 95%
3-Fluoro-4-nitrobenzoyl chloride
Formula: C₇H₃ClFNO₃
Boiling Pt: 109…110 °C (2 mm Hg)
Melting Pt: 23…25 °C
Storage Temperature: Ambient
MDL Number: MFCD03094235
CAS Number: 157665-51-5
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR