5-Trifluoromethyl-2H-pyrazol-3-ol hydrochloride
Supplier: Apollo Scientific
PC446037-1G
PC446037-500MG
APOSPC446037-1GEA
317
EUR
APOSPC446037-1G
APOSPC446037-500MG
5-Trifluoromethyl-2H-pyrazol-3-ol hydrochloride
5-Trifluoromethyl-2H-pyrazol-3-ol hydrochloride
Formula:
C₄H₃F₃N₂O·ClH MW: 188,536 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11845723 CAS Number: 866496-24-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...